Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADRA1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195843.1 | NP_001182772.1 | 1 - 2 | 887 | 198 | |
XM_005635607.3 | XP_005635664.1 | 3 - 4 | 1944 | 198 | ||
XM_022409542.1 | XP_022265250.1 | 2 - 3 | 2194 | 198 | ||
XM_022409544.1 | XP_022265252.1 | 2 - 3 | 2198 | 198 | ||
XM_022409545.1 | XP_022265253.1 | 2 - 3 | 2196 | 198 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Canis familiaris Probe spans exons |