Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIGD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005629355.2 | XP_005629412.1 | 2 | 1068 | 133 | |
XM_005629358.2 | XP_005629415.1 | 3 | 1154 | 133 | ||
XM_022403520.1 | XP_022259228.1 | 3 | 1157 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |