Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005623217.3 | XP_005623274.1 | 3 - 4 | 4631 | 178 | |
XM_547725.5 | XP_547725.2 | 3 - 4 | 4628 | 175 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |