Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005631789.3 | XP_005631846.1 | 583 | 128 | ||
XM_022405906.1 | XP_022261614.1 | 1203 | 128 | |||
XM_022405908.1 | XP_022261616.1 | 1130 | 128 | |||
XM_022405909.1 | XP_022261617.1 | 976 | 128 | |||
XM_022405910.1 | XP_022261618.1 | 1059 | 128 | |||
XM_022405911.1 | XP_022261619.1 | 972 | 128 | |||
XM_022405912.1 | XP_022261620.1 | 958 | 128 | |||
XM_022405913.1 | XP_022261621.1 | 950 | 89 | |||
XM_022405914.1 | XP_022261622.1 | 937 | 128 | |||
XM_022405915.1 | XP_022261623.1 | 930 | 89 | |||
XM_022405916.1 | XP_022261624.1 | 916 | 89 | |||
XM_022405917.1 | XP_022261625.1 | 594 | 128 | |||
XM_022405919.1 | XP_022261627.1 | 896 | 89 | |||
XM_022405920.1 | XP_022261628.1 | 560 | 128 | |||
XM_022405921.1 | XP_022261629.1 | 467 | 128 | |||
XM_022405922.1 | XP_022261630.1 | 547 | 128 | |||
XM_022405923.1 | XP_022261631.1 | 432 | 128 |
Set Membership: |
Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Recommended Assay Canis familiaris |