Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022423953.1 | XP_022279661.1 | 837 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Canis familiaris |