Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048138.1 | NP_001041603.1 | 3 - 4 | 361 | 84 | |
GenBank mRNA | DQ845341.1 | - | 3 - 4 | 361 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Probe spans exons |