Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST8SIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097548.1 | NP_001091017.1 | 4 - 5 | 714 | 91 | |
XM_005618263.3 | XP_005618320.1 | 5 - 6 | 908 | 91 | ||
GenBank mRNA | AM422132.1 | - | 4 - 5 | 714 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |