Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097551.1 | NP_001091020.1 | 7 - 8 | 593 | 95 | |
XM_022426505.1 | XP_022282213.1 | 7 - 8 | 598 | 95 | ||
GenBank mRNA | EF121758.1 | - | 7 - 8 | 593 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |