Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT2B31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003381.1 | NP_001003381.1 | 3 - 4 | 998 | 125 | |
GenBank mRNA | AY135176.1 | - | 3 - 4 | 998 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Inventoried Probe spans exons |