Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205190.2 | NP_001192119.1 | 2 - 3 | 516 | 101 | |
XM_005633260.3 | XP_005633317.1 | 3 - 4 | 667 | 101 | ||
XM_014122406.2 | XP_013977881.1 | 3 - 4 | 662 | 101 | ||
XM_014122409.2 | XP_013977884.1 | 3 - 4 | 662 | 101 | ||
XM_022407407.1 | XP_022263115.1 | 4 - 5 | 824 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Canis familiaris Probe spans exons |