Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDNRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010943.2 | NP_001010943.2 | 6 - 7 | 1214 | 105 | |
XM_005633820.3 | XP_005633877.1 | 7 - 8 | 1591 | 105 | ||
GenBank mRNA | AF276427.1 | - | 6 - 7 | 1214 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Canis familiaris Inventoried Probe spans exons |