Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS4X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252042.1 | NP_001238971.1 | 2 - 3 | 116 | 83 | |
XM_022415495.1 | XP_022271203.1 | 2 - 3 | 345 | 83 | ||
GenBank mRNA | DN358548.1 | - | 2 - 3 | 116 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |