Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252267.1 | NP_001239196.1 | 2 - 3 | 176 | 100 | |
GenBank mRNA | GR892513.1 | - | 2 - 3 | 147 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Canis familiaris Probe spans exons |