Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIVM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252348.1 | NP_001239277.1 | 3 - 4 | 218 | 88 | |
XM_005633838.3 | XP_005633895.1 | 6 - 7 | 1162 | 88 | ||
XM_005633839.3 | XP_005633896.1 | 6 - 7 | 1162 | 88 | ||
XM_014106255.2 | XP_013961730.1 | 6 - 7 | 1175 | 88 | ||
XM_022408005.1 | XP_022263713.1 | 6 - 7 | 1162 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Canis familiaris Probe spans exons |