Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpv17l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643374.1 | XP_007641564.1 | 4 - 5 | 511 | 98 | |
GenBank mRNA | JI876214.1 | - | 2 - 3 | 103 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |