Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ilk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246814.2 | NP_001233743.1 | 8 - 9 | 880 | 71 | |
GenBank mRNA | AB443452.1 | - | 8 - 9 | 881 | 71 | |
JI887863.1 | - | 8 - 9 | 935 | 71 | ||
JP050977.1 | - | 8 - 9 | 936 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons |