Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650202.1 | XP_007648392.1 | 5 - 6 | 551 | 85 | |
GenBank mRNA | JI862790.1 | - | 4 - 5 | 352 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase Detect Genomic DNA Recommended Assay Probe spans exons |