Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649414.1 | XP_007647604.1 | 2 - 3 | 5744 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Probe spans exons |