Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500049.2 | XP_003500097.1 | 9 - 10 | 2680 | 79 | |
XM_003500050.2 | XP_003500098.1 | 8 - 9 | 2416 | 79 | ||
XM_003500051.2 | XP_003500099.1 | 9 - 10 | 1737 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |