Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504685.2 | XP_003504733.2 | 4 - 5 | 492 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |