Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246784.1 | NP_001233713.1 | 3 - 4 | 249 | 96 | |
GenBank mRNA | AY302220.1 | - | 3 - 4 | 249 | 96 | |
JI885704.1 | - | 4 - 5 | 378 | 96 | ||
JP050082.1 | - | 4 - 5 | 377 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |