Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246825.2 | NP_001233754.1 | 1 - 2 | 510 | 68 | |
GenBank mRNA | JI890554.1 | - | 1 - 2 | 582 | 68 | |
U49649.1 | - | 1 - 2 | 510 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Endogenous Controls Detect Genomic DNA Recommended Assay Probe spans exons |