Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap3m1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244405.1 | NP_001231334.1 | 3 - 4 | 584 | 112 | |
XM_007652612.1 | XP_007650802.1 | 4 - 5 | 621 | 112 | ||
XM_007652618.1 | XP_007650808.1 | 4 - 5 | 751 | 112 | ||
GenBank mRNA | AY346109.1 | - | 4 - 5 | 851 | 112 | |
JI885336.1 | - | 4 - 5 | 942 | 112 | ||
JP040800.1 | - | 4 - 5 | 624 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |