Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244250.1 | NP_001231179.1 | 1 - 2 | 413 | 70 | |
GenBank mRNA | JI870076.1 | - | 1 - 2 | 402 | 70 | |
JP043410.1 | - | 1 - 2 | 402 | 70 | ||
Z46801.1 | - | 1 - 2 | 413 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |