Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244250.1 | NP_001231179.1 | 3 - 4 | 804 | 76 | |
GenBank mRNA | Z46801.1 | - | 3 - 4 | 804 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |