Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500330.2 | XP_003500378.1 | 2 | 660 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Both primers and probe map within a single exon |