Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc85c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503715.2 | XP_003503763.1 | 5 - 6 | 446 | 59 | |
GenBank mRNA | JP064708.1 | - | 2 - 3 | 138 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |