Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cinp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498777.2 | XP_003498825.1 | 2 - 3 | 222 | 77 | |
XM_007632581.1 | XP_007630771.1 | 2 - 3 | 188 | 77 | ||
GenBank mRNA | JI873083.1 | - | 2 - 3 | 224 | 77 | |
JP049833.1 | - | 2 - 3 | 224 | 77 |