Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501236.1 | XP_003501284.1 | 1 - 2 | 625 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Probe spans exons |