Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fosl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496572.2 | XP_003496620.1 | 1 - 2 | 864 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |