Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515375.1 | XP_003515423.1 | 5 - 6 | 919 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Transporter 3' Most Detect Genomic DNA Probe spans exons |