Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp8ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641106.1 | XP_007639296.1 | 2 - 3 | 292 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Apoptosis Recommended Assay Probe spans exons |