Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zscan30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641539.1 | XP_007639729.1 | 1 - 2 | 2213 | 96 | |
XM_007641540.1 | XP_007639730.1 | 2 - 3 | 623 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |