Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100751713 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498025.1 | XP_003498073.1 | 4 - 5 | 354 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |