Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plet1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498539.2 | XP_003498587.2 | 2 - 3 | 492 | 79 | |
GenBank mRNA | JI870244.1 | - | 2 - 3 | 458 | 79 | |
JP063116.1 | - | 2 - 3 | 456 | 79 |