Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510706.2 | XP_003510754.1 | 1 - 2 | 675 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Probe spans exons |