Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512971.2 | XP_003513019.2 | 3 - 4 | 890 | 109 | |
XM_007653345.1 | XP_007651535.1 | 3 - 4 | 872 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |