Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rprd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641549.1 | XP_007639739.1 | 4 - 5 | 365 | 85 | |
GenBank mRNA | JI867564.1 | - | 569 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |