Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pidd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509765.2 | XP_003509813.1 | 10 - 11 | 2004 | 65 | |
XM_007650419.1 | XP_007648609.1 | 10 - 11 | 1909 | 65 | ||
XM_007650420.1 | XP_007648610.1 | 10 - 11 | 2000 | 65 | ||
XM_007650421.1 | XP_007648611.1 | 10 - 11 | 2048 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Probe spans exons |