Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498894.1 | XP_003498942.1 | 1 - 2 | 1082 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Probe spans exons |