Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498894.1 | XP_003498942.1 | 1 | 917 | 93 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Both primers and probe map within a single exon |