Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slitrk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651774.1 | XP_007649964.1 | 2 - 3 | 2611 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Kinase Recommended Assay Probe spans exons |