Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slitrk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651774.1 | XP_007649964.1 | 3 - 4 | 2765 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Protein Kinase 3' Most Probe spans exons |