Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509500.1 | XP_003509548.1 | 4 - 5 | 655 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Ion Channel 3' Most Probe spans exons |