Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640764.1 | XP_007638954.1 | 3 - 4 | 431 | 85 | |
GenBank mRNA | JI893406.1 | - | 2 - 3 | 378 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |