Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsen54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495697.2 | XP_003495745.1 | 11 - 12 | 1668 | 90 | |
GenBank mRNA | JI891630.1 | - | 2 - 3 | 181 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |