Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503373.2 | XP_003503421.1 | 3 - 4 | 311 | 62 | |
XM_007643944.1 | XP_007642134.1 | 3 - 4 | 301 | 62 | ||
XM_007643945.1 | XP_007642135.1 | 3 - 4 | 303 | 62 | ||
GenBank mRNA | JI883570.1 | - | 420 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons |