Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Extl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003497962.2 | XP_003498010.1 | 4 - 5 | 3198 | 65 | |
XM_007620249.1 | XP_007618439.1 | 5 - 6 | 3249 | 65 | ||
XM_007620255.1 | XP_007618445.1 | 4 - 5 | 3141 | 65 | ||
GenBank mRNA | JI877152.1 | - | 2 - 3 | 331 | 65 | |
JP062845.1 | - | 2 - 3 | 331 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |