Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502004.2 | XP_003502052.1 | 5 - 6 | 617 | 98 | |
GenBank mRNA | JI890993.1 | - | 1 - 2 | 179 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |