Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Irx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511366.1 | XP_003511414.1 | 3 - 4 | 1175 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons |